Skip to Content
Merck
All Photos(1)

Documents

93738

Supelco

γ-Decalactone

analytical standard

Synonym(s):

γ-Decanolactone, 4-Hydroxydecanoic acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.447-1.451
n20/D 1.449 (lit.)

density

0.948 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCC1CCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-5-6-9-7-8-10(11)12-9/h9H,2-8H2,1H3

InChI key

IFYYFLINQYPWGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Decalactone is a flavor and fragrance compound, which finds applications in food and beverage industry.

Application

γ-Decalactone may be used as an analytical reference standard for the quantification of the analyte in fruit beverages using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On-line reversed?phase liquid chromatography-gas chromatography coupled to mass spectrometry for enantiomeric analysis of chiral compounds in fruit beverages
Caja.MM, et al.
Journal of Chromatography A, 1054(1-2), 81-85 (2004)
Characterisation of volatile compounds of fruit juices and nectars by HS/SPME and GC/MS
Riu-Aumatell.M
Food Chemistry, 87(4), 627-637 (2004)
Quantitative analysis of ??and ??lactones in wines using gas chromatography with selective tandem mass spectrometric detection
Langen J, et al.
Rapid Communications in Mass Spectrometry, 27(24), 2751-2759 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service