Skip to Content
Merck
All Photos(1)

Key Documents

G1007617

HPLC Analysis of Atorvastatin and Related Compound E on Kromasil® 5-AmyCoat Chiral

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Kromasil 5-AmyCoat, 25 cm x 4.6 mm, 5 μm particles (K08670348)
mobile phase: 84:16 hexane: ethanol with 0.1% TFA
flow rate: 1 mL/min
pressure: 1117 psi (77 bar)
column temp.: 25 °C
detector: UV, 254 nm
injection: 20 μL
sample: 50 μg/mL atorva

suitability

application for HPLC

application(s)

pharmaceutical (small molecule)

Analysis Note

Atorvastatin calcium also known as HMG-CoA reductase inhibitors in a class of lipid-lowering medications that reduces levels of “bad” cholesterol (low-density lipoprotein, or LDL) and triglycerides in the blood and raise “good” cholesterol (high-density lipoprotein, or HDL). The enantiomer of atorvastatin (related compound E) could qualitatively and quantitatively differ in their activities, in pharmacokinetic behavior or pharmacodynamics. The application exhibits baseline resolution under 9 minutes of a racemic mixture of atorvastatin calcium and related compound E on a 5.0 μm Kromasil 5-AmyCoat Chiral.

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Analyte

Description

    Atorvastatin related compound E (PHR1872)
    Atorvastatin Calcium, Pharmaceutical Secondary Standard; Certified Reference Material

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service