Skip to Content
Merck
All Photos(1)

Key Documents

32293

Sigma-Aldrich

Hexane

≥99% (GC), ACS reagent, suitable for GC/MS, puriss. p.a., reag. Ph. Eur.

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/MS: suitable
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

GC/MS: suitable

impurities

≤0.0001% thiophene
≤0.001% non-volatile matter
≤0.002% free acid (as CH3COOH)
≤0.005% S-compounds (as S)
≤0.01% aromatic substances (as C6H6)
≤0.01% water (Karl Fischer)

refractive index

n20/D 1.3745-1.3755
n20/D 1.375 (lit.)

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCCCCC

suitability

passes test for reaction against H2SO4

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexane is an alkane hydrocarbon having 5 structural isomeric forms. It is a colorless, combustible liquid. It is one of the essential components of petroleum ether. It can be obtained from petroleum by distillation. It can also be produced by electrolysis of potassium butyrate. It is widely employed organic solvent for the dissolution of fats and oils.

Application

Hexane has been used in the preparation of graft-copolymer chains of poly-(dimethylsiloxane)-g-poly(methyl methacrylate). It has been employed for the GC/MS characterization of essential oils extracted from various origins.

Analysis Note

boiling range 67-69°C min. 95%; density (D 20/20) 0.659-0.663

Other Notes

The article number 32293-4X2.5L will be discontinued. Please order the single bottle 32293-2.5L which is physically identical with the same exact specifications.
The article number 32293-4X2.5L-M will be discontinued. Please order the single bottle 32293-2.5L-M which is physically identical with the same exact specifications.
The article number 32293-6X1L will be discontinued. Please order the single bottle 32293-1L which is physically identical with the same exact specifications.
The article number 32293-6X1L-M will be discontinued. Please order the single bottle 32293-1L-M which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jonas B Nielsen et al.
Nature communications, 11(1), 6417-6417 (2020-12-20)
Pharmaceutical drugs targeting dyslipidemia and cardiovascular disease (CVD) may increase the risk of fatty liver disease and other metabolic disorders. To identify potential novel CVD drug targets without these adverse effects, we perform genome-wide analyses of participants in the HUNT
Comparative evaluation of 11 essential oils of different origin as functional antioxidants, antiradicals and antimicrobials in foods.
Sacchetti G, et al.
Food Chemistry, 91(4), 621-632 (2005)
Ali Mohraz et al.
Langmuir : the ACS journal of surfaces and colloids, 21(12), 5298-5306 (2005-06-01)
The development of model materials and image processing methods to directly visualize and quantify colloidal rod assembly by means of confocal laser scanning microscopy (CLSM) is reported. Monodisperse fluorescent colloidal rods are prepared by the uniaxial extensional deformation of sterically
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of
Alexandre S Cristino et al.
Nature communications, 5, 5529-5529 (2014-11-21)
Increasing evidence suggests small non-coding RNAs (ncRNAs) such as microRNAs (miRNAs) control levels of mRNA expression during experience-related remodelling of the brain. Here we use an associative olfactory learning paradigm in the honeybee Apis mellifera to examine gene expression changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service