Skip to Content
Merck
All Photos(1)

Documents

G5537

Sigma-Aldrich

Glyceraldehyde-3-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae)

lyophilized powder, 70-140 units/mg protein

Synonym(s):

D-Glyceraldehyde 3-phosphate:NAD+ oxidoreductase (phosphorylating), GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

Quality Level

specific activity

70-140 units/mg protein

composition

Protein, 30-70% biuret

foreign activity

3-Phosphoglyceric phosphokinase ≤0.05%

storage temp.

−20°C

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase catalyzes the conversion of glyceraldehyde-3-phosphate to 1,3-bisphosphoglycerate as part of glycolysis. It has also been shown to have roles in initiation of apoptosis, transcription activation and the shuttling of ER to Golgi vesicles<<<,New>>>.

Unit Definition

One unit will reduce 1.0 μmole of 3-phosphoglycerate to D-glyceraldehyde 3-phosphate per min in a coupled system with 3-phosphoglyceric phosphokinase at pH 7.6 at 25 °C.

Physical form

Lyophilized, sulfate-free powder stabilized with trehalose, citrate, and DTT. Useful for systems requiring low sulfate.

inhibitor

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Tong et al.
The Journal of biological chemistry, 284(17), 11256-11266 (2009-03-03)
Although the classical redox functions of co-enzyme NAD(+) are firmly established in metabolism, there are numerous enzymes that catalyze cleavage of NAD(+) to yield free ADP-ribose (ADPr) or related metabolites, whose functions remain largely unknown. Here we show that the
A Tarze et al.
Oncogene, 26(18), 2606-2620 (2006-10-31)
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a pleiotropic enzyme that is overexpressed in apoptosis and in several human chronic pathologies. Here, we report that the protein accumulates in mitochondria during apoptosis, and induces the pro-apoptotic mitochondrial membrane permeabilization, a decisive event of
Bing Fang et al.
Biochimie, 95(2), 251-257 (2012-09-27)
siRNA is promising in anti-tumor therapy. The main challenge is lack of tumor-specific intracellular delivery. In this study, a 6 amino acids peptide (A1) with high affinity for vascular endothelial growth factor receptor-1 (VEGFR1) was conjugated with a cell penetrating
Pablo Moreno-Acosta et al.
Anticancer research, 32(10), 4319-4325 (2012-10-13)
The aim of this study was to evaluate the predictive utility of Insulin-like growth factor-1 receptor (IGF1R), IGF1, IGF2, Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) and of hemoglobin levels for tumor response to exclusive radiotherapy, in patients with locally advanced Human papillomavirus
Tina C Albershardt et al.
Journal of immunological methods, 384(1-2), 196-199 (2012-08-14)
Normalization to a reference gene is the method of choice for quantitative PCR analysis. The stability of reference genes is critical for accurate gene expression analysis, as significant variations in reference gene expression can alter experimental results and conclusions. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service