Key Documents
HS15092ZP
Standard Fine Tip Sharpie® Marker
black
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
material
black
packaging
pack of 12 ea
manufacturer/tradename
Heathrow Scientific HS15092ZP
General description
Standard Fine Tip Sharpie® Marker, Black
Legal Information
Sharpie is a registered trademark of Sanford, LP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. is this pen alcohol resistant?
    1 answer
    1. This product is not resistant to alcohol. It is resistant to fading and water exposure.
      Helpful?