Skip to Content
Merck
All Photos(1)

Key Documents

EMT6/AR10.0

96042326, mouse unknown/unspecified, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

EMT6/AR10.0, 96042326

biological source

mouse unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse mammary, drug-resistant

Cell Line Description

The drug resistant cell line EMT6/AR10.0 was derived from the mouse mammary tumour cell line EMT6/P (Sigma Catalogue number. 96042344) by continuous growth in increasing concentrations of doxorubicin. It was reported to show reduced intracellular drug accumulation and hyperexpression of P-glycoprotein. It is recommended to culture the cells without drug after resuscitation until the first passage.

Application

Drug-resistance studies

Culture Medium

MEM(EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA)+ 10.0μg/ml doxorubicin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin or trypsin/EDTA; 5%CO2; 37ºC; resuscitate in drug-free media, add doxorubicin after first passage

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service