Skip to Content
Merck
All Photos(1)

Key Documents

NIST968F

Fat-Soluble Vitamins in Frozen Human Serum

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

biological source

human

form

liquid

packaging

vial of 2 × 1 mL

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

solubility

water: slightly soluble

application(s)

clinical testing

storage temp.

−70°C

General description

A unit of NIST SRM 968f consists of two vials, each containing approximately 1 mL of frozen human serum at one of two different concentration levels.

NIST968F_Cert

NIST968F_SDS

Application

This fat-soluble vitamin standard reference material (SRM) is intended for use in validating methods for determining fat-soluble vitamins in human serum and plasma. It has also been used for qualifying control materials produced in-house and analyzed using those methods.

Other Notes

  • Instructions on safety measures, usage, storage, and handling are available in the certificate.
  • Details on transportation, disposal, and biomaterials are available in MSDS.
Certified for the analytes listed below.
Total Retinol, α-Tocopherol, α+α-Tocopherol

Reference/Informational value is provided for the following analytes.
Total α-Carotene, Trans-α-Carotene, Cis-α-Carotene, Total α-Carotene, Total Lycopene, Trans-Lycopene, Total α Cryptoxanthin, Total Lutein, Total Zeaxanthin, Total Lutein+Zeaxanthin, Retinyl Palmitate, Coenzyme Q10, Phylloquinone, 25-Hydroxyvitamin D
Vitamin D Metabolite, 25-Hydroxyvitamin D2, 25-Hydroxyvitamin D3, 3-epi-25-Hydroxyvitamin D3, Serum Density

See certificate for values and more details at nist.gov/SRM.

Preparation Note

  • The vials should be stored in the dark at a temperature between –70 °C.
  • Before use, the vials must be gently mixed and allowed to thaw at room temperature.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Zhang et al.
Analytical methods : advancing methods and applications, 14(25), 2511-2521 (2022-06-16)
Several observational studies have reported associations between low levels of fat-soluble vitamins (A, D and E) and the incidence of pneumonia. Whether infection affects or negatively regulates serum vitamin levels remains controversial. Our aims were to develop and validate a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service