Skip to Content
Merck
All Photos(1)

Key Documents

NIST2969

Vitamin D Metabolites in Frozen Human Serum (Total 25-Hydroxyvitamin D Low Level)

NIST®SRM®

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
NACRES:
NA.77

packaging

pkg of 2 × 1 mL

manufacturer/tradename

NIST®

application(s)

clinical testing

format

multi-component solution

storage temp.

−70°C

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service