Skip to Content
Merck
All Photos(4)

Documents

44290

Sigma-Aldrich

AmberChrom 1X2 chloride form

50-100 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

description

strongly basic

Quality Level

form

beads

parameter

66 °C OH- form max. temp.
99 °C Cl- form max. temp.

moisture

65-75%

loss

~70% loss on drying, 110 °C

matrix

styrene-divinylbenzene (gel)

matrix active group

trimethylbenzylammonium functional group

particle size

50-100 mesh

operating pH

0-14

capacity

0.7 meq/mL by wetted bed volume

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

AmberChrom 1X2 (formerly Dowex 1X2) is an anion exchange resin.

Application

AmberChrom 1X2 chloride form has been used in the purification step for the synthesis of methyl-esterified sialic acid analogs bearing a diazirine photocrosslinker on the N-acyl (C-5) side chain 5-(SiaDAz).
AmberChrom 1X2 chloride form is a strongly basic (type I) anion exchange resin for food & drug processing. it has been used to characterize a dual emission sensing luminescence material for water-dissolved oxygen sensing. AmberChrom ion exchange resin has also been used to demonstrate decontamination of (99g) Tc from spent 99Mo waste solution and recovery of (99g)Tc for research use.
Strongly basic (type I) anion exchange resin for food & drug processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocrosslinking of glycoconjugates using metabolically incorporated diazirine-containing sugars
Bond MR, et al.
Nature Protocols, 4(7), 1044-1044 (2009)
Sankha Chattopadhyay et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 66(10), 1295-1299 (2008-04-05)
Molybdenum-99 of low-to-medium specific activity, 7.4-14.8 GBq/g, obtained by the (98)Mo(n,gamma)(99)Mo reaction in research reactors requires a large alumina column to adsorb the molybdenum since the capacity of alumina to adsorb Mo is limited (20 mg Mo/g of alumina). The
Hannes Hochreiner et al.
Talanta, 66(3), 611-618 (2008-10-31)
The characterization of a dual emission sensing luminescence material for water-dissolved oxygen sensing is presented in this paper. The oxygen-sensitive material is based on a dual-emitting luminescent molecule immobilized onto an adequate solid support. The metal chelate formed between the
Sankha Chattopadhyay et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 66(12), 1814-1817 (2008-08-16)
A simple separation technique of (99m)Tc from Na(2)[(99)Mo]MoO(4) in sodium hydroxide solution obtained from the (98)Mo(n,gamma)(99)Mo reaction is described. Low to medium specific activity (99)Mo-molybdate solution of 7.4-18.5GBq (200-500mCi) in sodium hydroxide was passed through a tiny Dowex-1 column (25mg)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service