Skip to Content
Merck
All Photos(1)

Key Documents

13547

Supelco

Diaion® CR11 sodium form

pkg of 1000 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

description

weakly acidic cation exchange resin

form

beads

packaging

pkg of 1000 g

parameter

120 °C Na+ form max. temp.
80 °C H+ form max. temp.

moisture

55-65%

technique(s)

LPLC: suitable

matrix

crosslinked polystyrene (highly porous)

matrix active group

iminodiacetic acid functional group

particle size

16-45 mesh

operating pH

4-10

capacity

0.35 meq/mL by wetted bed volume (calcium)
1.2 meq/g by dry weight (calcium)

separation technique

affinity

General description

Diaion CR11 is chelating resin with iminodiacetic acid groups. It is considered effective in the removal of chromium trivalent from aqueous solutions. This resin in the Na+ form at 50°C is seen to exhibit high sorption capacity.

Application

Diaion CR11 was used to prepare phosphate buffer, required to measure rate constants of the reactions of hypochlorite with L-Arg and taurine by stopped-flow spectrophotometry.
Chelating resin for metal recovery, wastewater treatment, brine purification, etc.

Legal Information

Diaion is a registered trademark of Mitsubishi Chemical Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia A Cavaco et al.
Journal of hazardous materials, 144(3), 634-638 (2007-03-06)
Effluent discharged from the chromium electroplating industry contains a large number of metals, including chromium, copper, nickel, zinc, manganese and lead. The ion exchange process is an alternative technique for application in the treatment of industrial wastewater containing heavy metals
Sofia A Cavaco et al.
Journal of hazardous materials, 169(1-3), 516-523 (2009-05-02)
In this study two chelating resins containing iminodiacetic acid groups (Amberlite IRC 748 and Diaion CR 11) and a chelating resin based on sulfonic and diphosphonic acid groups (Diphonix) were investigated in order to separate Cr(III) from industrial effluents produced
C Zhang et al.
The Journal of biological chemistry, 276(29), 27159-27165 (2001-05-26)
The myeloperoxidase-derived oxidant hypochlorous acid (HOCl) is thought to contribute to endothelial dysfunction, but the mechanisms underlying this inhibitory effect are unknown. The present study tested the hypothesis that HOCl and L-arginine (L-Arg) react to form novel compounds that adversely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service