Key Documents
Z232386
Pouring rings for Duran® laboratory bottles
GL 32 neck thread, red melamine resin
Synonym(s):
DURAN pouring ring, ETFE, GL32, high temperature, pouring ring
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
red melamine resin
manufacturer/tradename
DURAN®
Schott
parameter
-100-180 °C temp. stability
fitting
GL 32 neck thread
General description
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
Features and Benefits
Optional pouring rings prevent drips when pouring from Duran bottles.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)