Key Documents
Z232378
Pouring rings for Duran® laboratory bottles
GL 32 neck thread, blue polypropylene
Synonym(s):
pouring ring
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
blue polypropylene
manufacturer/tradename
DURAN®
Schott
parameter
-40-140 °C temp. stability
fitting
GL 32 neck thread
General description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Features and Benefits
Optional pouring rings prevent drips when pouring from Duran bottles.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)