Skip to Content
Merck
All Photos(1)

Key Documents

912514

Sigma-Aldrich

N-Nitrosaccharin

Synonym(s):

2-Nitrobenzo[d]isothiazolone 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4N2O5S
CAS Number:
Molecular Weight:
228.18
UNSPSC Code:
12352200

form

powder

availability

available only in USA

storage temp.

−20°C

Application

2-Nitrobenzo[d]isothiazolone 1,1-dioxide is easy-to-handle, bench stable nitration reagent that enables access to nitro(hetero)arenes under mild reaction conditions and in high efficiency.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roxan Calvo et al.
Nature communications, 10(1), 3410-3410 (2019-08-01)
Nitroaromatics and nitroheteroaromatics serve as key building blocks and intermediates in synthesis, and form the core scaffold of a vast number of materials, dyes, explosives, agrochemicals and pharmaceuticals. However, their synthesis relies on harsh methodologies involving excess mineral acids, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service