Skip to Content
Merck
All Photos(1)

Key Documents

329908

Sigma-Aldrich

Diazald®-(N-methyl-d3)

98 atom % D, 97% (CP)

Synonym(s):

4-Methyl-N-(methyl-d3)-N-nitrosobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2N(CD3)NO
CAS Number:
Molecular Weight:
217.26
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

solid

mp

60-63 °C (lit.)

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])N(N=O)S(=O)(=O)c1ccc(C)cc1

InChI

1S/C8H10N2O3S/c1-7-3-5-8(6-4-7)14(12,13)10(2)9-11/h3-6H,1-2H3/i2D3

InChI key

FFKZOUIEAHOBHW-BMSJAHLVSA-N

Application

Reagent for preparing diazomethane of higher isotopic purity than is available using the Deutero-Diazald prep set.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service