Skip to Content
Merck
All Photos(1)

Key Documents

SRP3057

Sigma-Aldrich

ICAM-I human

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CD54 antigen, Intercellular adhesion molecule 1, major group rhinovirus receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

sterility

sterile

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

25 ng/mL

mol wt

72.0-80.0 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ICAM1(3383)

General description

ICAM1 (intercellular adhesion molecule 1) was initially identified as a ligand for the β2 integrin lymphocyte function-associated antigen (LFA)-1. It is a glycoprotein with a 90,000-110,000Mr molecular weight, and shows wide range of tissue expression, including tissue macrophages and dendritic cells. It also shows expression on non-hematopoietic cells such as vascular endothelial cells, epithelial cells of thymus and mucosa, and dermal fibroblasts. It is a member of the ICAM family, which also include ICAM-2, -3, -4 and -5.
Recombinant human ICAM-1 is a 49.5 kDa glycoprotein comprising the extracellular domain (453 amino acid residues) of ICAM-1. Monomeric glycosylated ICAM-1 migrates at an apparent molecular weight of approximately 72.0-80.0 kDa by SDS-PAGE analysis under reducing conditions.

Biochem/physiol Actions

The expression of ICAM1 (intercellular adhesion molecule 1) is strongly increased by IL-1 (interleukin) and interferon (IFN)-γ. It is expressed on lymphoid cells where it is required for phorbol ester-induced aggregation. It participates in LFA-1 (lymphocyte function-associated antigen)-dependent NK (natural killer) cell stimulation through its attachment to actin cytoskeleton through α-actinin or ezrin. It also acts as the receptor for rhinovirus, a common cold virus and Plasmodium falciparum. Soluble ICAM-1 levels are elevated in pregnant women with gestational diabetes mellitus (GDM), which is an outcome of endothelial cell activation and dysfunction. Meta-analysis of case-control studies show that rs1799969 polymorphism in ICAM-1 gene is linked with increased susceptibility to cancer.

Sequence

QTSVSPSKVI LPRGGSVLVT CSTSCDQPKL LGIETPLPKK ELLLPGNNRK VYELSNVQED SQPMCYSNCP DGQSTAKTFL TVYWTPERVE LAPLPSWQPV GKNLTLRCQV EGGAPRANLT VVLLRGEKEL KREPAVGEPA EVTTTVLVRR DHHGANFSCR TELDLRPQGL ELFENTSAPY QLQTFVLPAT PPQLVSPRVL EVDTQGTVVC SLDGLFPVSE AQVHLALGDQ RLNPTVTYGN DSFSAKASVS VTAEDEGTQR LTCAVILGNQ SQETLQTVTI YSFPAPNVIL TKPEVSEGTE VTVKCEAHPR AKVTLNGVPA QPLGPRAQLL LKATPEDNGR SFSCSATLEV AGQLIHKNQT RELRVLYGPR LDERDCPGNW TWPENSQQTP MCQAWGNPLP ELKCLKDGTF PLPIGESVTV TRDLEGTYLC RARSTQGEVT RKVTVNVLSP RYE

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ICAM-1 (CD54): a counter-receptor for Mac-1 (CD11b/CD18).
Diamond MS et al
The Journal of Cell Biology, 111(6 Pt 2), 3129-3139 (1990)
Intercellular Adhesion Molecule and Endogenous NOS Inhibitor: Asymmetric Dimethylarginine in Pregnant Women with Gestational Diabetes Mellitus.
Poniedzialek-Czajkowska E et al
Journal of Diabetes Research, 2016, 1342643-1342643 (2016)
ICAM-1: getting a grip on leukocyte adhesion.
Long EO
Journal of Immunology, 186(9), 5021-5023 (2011)
Charlotte Lawson et al.
Pharmacological reports : PR, 61(1), 22-32 (2009-03-25)
Intercellular adhesion molecule-1 (ICAM-1; CD54) is a 90 kDa member of the immunoglobulin (Ig) superfamily and is critical for the firm arrest and transmigration of leukocytes out of blood vessels and into tissues. ICAM-1 is constitutively present on endothelial cells
Association of Polymorphisms in Intercellular Adhesion Molecule 1 (ICAM-1) Gene with Cancer Susceptibility: A Meta-Analysis of 14 Case-Control Studies.
Zhang X et al
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 569-579 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service