Skip to Content
Merck
All Photos(1)

Key Documents

SRP3182

Sigma-Aldrich

VEGF human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Folliculostellate cell-derived growth factor, Glioma-derived endothelial cell mitogen, VPF, Vascular Endothelial Growth Factor-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-8.0 ng/mL

mol wt

38.2 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGFA(7422)

General description

VEGF (vascular endothelial growth factor) signals through the three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant human VEGF is a 38.2kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.

Application

VEGF human has been used as a chemoattractant for HUVECs (human umbilical vein endothelial cells) migration assay.

Biochem/physiol Actions

VEGF (vascular endothelial growth factor) is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intraocular neovascular syndromes. Polymorphism in VEGF at C(-634)G is linked with diabetic retinopathy in type 2 diabetes.

Sequence

APMAEGGGQN HHEVVKFMDV YQRSYCHPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCGGC CNDEGLECVP TEESNITMQI MRIKPHQGQH IGEMSFLQHN KCECRPKKDR ARQENPCGPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Horikawa et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(2), 587-599 (2016-07-13)
High VEGF expression in ovarian cancer is an unfavorable prognostic factor. However, the role of VEGF in tumor immunity remains unclear. Here, we examined the impact of VEGF on local immunity, including induction of myeloid-derived suppressor cells (MDSC), in ovarian
Vegf, Vegf-B, Vegf-C and their receptors KDR, FLT-1 and FLT-4 during the neoplastic progression of human colonic mucosa.
Andre T
International Journal of Cancer. Journal International Du Cancer, 86, 174-181 (2000)
Naoko Takubo et al.
Scientific reports, 9(1), 9304-9304 (2019-06-28)
Vascular endothelial cells (ECs) in angiogenesis exhibit inhomogeneous collective migration called "cell mixing", in which cells change their relative positions by overtaking each other. However, how such complex EC dynamics lead to the formation of highly ordered branching structures remains
M Clauss et al.
The Journal of experimental medicine, 172(6), 1535-1545 (1990-12-01)
Systemic infusion of low concentrations of tumor necrosis factor/cachectin (TNF) into mice that bear TNF-sensitive tumors leads to activation of coagulation, fibrin formation, and occlusive thrombosis exclusively within the tumor vascular bed. To identify mechanisms underlying the localization of this
Ca2+ influx through reverse mode Na+/Ca2+ exchange is critical for vascular endothelial growth factor-mediated extracellular signal-regulated kinase (ERK) 1/2 activation and angiogenic functions of human endothelial cells.
Andrikopoulos P
The Journal of Biological Chemistry, 286, 37919-37931 (2011)

Articles

Read review on VEGF signaling pathway (Vascular endothelial growth factor (VEGF)/VEGF receptor) and find related products.

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

See All

Protocols

Discover our collection of primary human hepatic sinusoidal endothelial cells, a protocol for thawing, plating, and growing HHSECs, and a recipe for our HHSEC maintenance media formulation.

Discover our collection of primary human hepatic sinusoidal endothelial cells, a protocol for thawing, plating, and growing HHSECs, and a recipe for our HHSEC maintenance media formulation.

Discover our collection of primary human hepatic sinusoidal endothelial cells, a protocol for thawing, plating, and growing HHSECs, and a recipe for our HHSEC maintenance media formulation.

Discover our collection of primary human hepatic sinusoidal endothelial cells, a protocol for thawing, plating, and growing HHSECs, and a recipe for our HHSEC maintenance media formulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service