Skip to Content
Merck
All Photos(1)

Key Documents

SAB4600054

Sigma-Aldrich

Anti-HA (Hemagglutinin) IgG, CF 488A antibody produced in rabbit

~1 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

CF 488A conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 1-10 μg/mL

fluorescence

λex 490 nm; λem 515 nm

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Influenza virus hemagglutinin (HA) precursor is encoded by the gene mapped on segment 4. The encoded protein is a 76kDa surface glycoprotein and it exists as trimers on the envelope.

Specificity

Binds HA epitope tag

Immunogen

proteins contatining the epitope tag sequence

Biochem/physiol Actions

Influenza virus hemagglutinin (HA) acts as a key immunogen in avian influenza virus (AIV) vaccines. Activated HA facilitates virus attachment to oligosaccharide receptors on host cell surface and is involved in penetration through fusion following conformational modifications in the endolysosomal compartment. 226 and 228 residues in hemagglutinin (HA) of the H3 subtype play a key role in host range restriction and receptor specificity. Influenza virus hemagglutinin (HA) is responsible for 1918 influenza pandemic.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 1 mg/mL bovine serum albumin.

Preparation Note

Protect from light.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A single amino acid substitution in 1918 influenza virus hemagglutinin changes receptor binding specificity.
Glaser L
Journal of Virology, 79, 11533-11536 (2005)
Visualization of Alternative Functional Configurations of Influenza Virus Hemagglutinin Facilitates Rapid Selection of Complementing Vaccines in Emergency Situations.
Metwally A and Yousif A
International Journal of Molecular Sciences, 18 (2017)
The role of influenza A virus hemagglutinin residues 226 and 228 in receptor specificity and host range restriction.
Vines A
Journal of Virology, 72, 7626-7631 (1998)
Anika Mann et al.
Scientific reports, 11(1), 8288-8288 (2021-04-17)
The dopamine D2 receptor (D2R) is the target of drugs used to treat the symptoms of Parkinson's disease and schizophrenia. The D2R is regulated through its interaction with and phosphorylation by G protein receptor kinases (GRKs) and interaction with arrestins.
Anika Mann et al.
Scientific reports, 10(1), 8585-8585 (2020-05-24)
The δ-opioid receptor (DOP) is an attractive pharmacological target due to its potent analgesic, anxiolytic and anti-depressant activity in chronic pain models. However, some but not all selective DOP agonists also produce severe adverse effects such as seizures. Thus, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service