Merck
  • Agonist-induced phosphorylation bar code and differential post-activation signaling of the delta opioid receptor revealed by phosphosite-specific antibodies.
Agonist-induced phosphorylation bar code and differential post-activation signaling of the delta opioid receptor revealed by phosphosite-specific antibodies.
Scientific reports (2020-05-24)
Anika Mann, Sophia Liebetrau, Marie Klima, Pooja Dasgupta, Dominique Massotte, Stefan Schulz
ABSTRACT
The δ-opioid receptor (DOP) is an attractive pharmacological target due to its potent analgesic, anxiolytic and anti-depressant activity in chronic pain models. However, some but not all selective DOP agonists also produce severe adverse effects such as seizures. Thus, the development of novel agonists requires a profound understanding of their effects on DOP phosphorylation, post-activation signaling and dephosphorylation. Here we show that agonist-induced DOP phosphorylation at threonine 361 (T361) and serine 363 (S363) proceeds with a temporal hierarchy, with S363 as primary site of phosphorylation. This phosphorylation is mediated by G protein-coupled receptor kinases 2 and 3 (GRK2/3) followed by DOP endocytosis and desensitization. DOP dephosphorylation occurs within minutes and is predominantly mediated by protein phosphatases (PP) 1α and 1β. A comparison of structurally diverse DOP agonists and clinically used opioids demonstrated high correlation between G protein-dependent signaling efficacies and receptor internalization. In vivo, DOP agonists induce receptor phosphorylation in a dose-dependent and agonist-selective manner that could be blocked by naltrexone in DOP-eGFP mice. Together, our studies provide novel tools and insights for ligand-activated DOP signaling in vitro and in vivo and suggest that DOP agonist efficacies may determine receptor post-activation signaling.
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
[D-Pen2,5]-Enkephalin hydrate, ≥95% (HPLC)
Sigma-Aldrich
[Met5]Enkephalin acetate salt hydrate, ≥95.0% (HPLC), powder
Sigma-Aldrich
Fentanyl citrate salt
Sigma-Aldrich
[D-Ala2]-Deltorphin II, ≥97% (HPLC)
Sigma-Aldrich
Anti-HA (Hemagglutinin) IgG, CF 488A antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody
Sigma-Aldrich
[D-Ala2, D-Leu5]-Enkephalin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
SNC80, ≥98% (HPLC), solid