Skip to Content
Merck
All Photos(1)

Documents

MISELADLA

Millipore

MilliSentials Lab Adaptable Labels

Synonym(s):

Custom lab labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
44102408
NACRES:
IA.22

packaging

roll of 1615 labels

Quality Level

roll size

1 in. core , Media Roll Size

General description

The MilliSentials Lab Adaptable labels have been custom designed and optimized for use throughout the laboratory. The labels are capable of withstanding temperatures from -196°C (liquid nitrogen) to 100°C (boiling water) and provide resistance to harsh chemical solvents (including ethanol, DMSO, xylene).
The labels have also been designed with perforations allowing one size to fit across a wide range of laboratory vessels including microcentrifuge tubes, cryovials, petri dishes, cell culture plates, and reagent bottles.
For use with the MilliSentials Lab Labeling System.

Application

Laboratory Sample Labeling

Features and Benefits

  • Single label size that can adapt across multiple laboratory vessels
  • Temperature resistance
  • Chemical solvent resistance

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.

MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.

MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.

MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service