Documents
MISELPRRI
Millipore
MilliSentials Lab Printer Ribbon
About This Item
UNSPSC Code:
44102408
NACRES:
IA.22
diam.
× 1 in. , Ribbon Core (Inner Diameter)
Quality Level
roll size
1 , ∼6400 labels printed from 1 roll of ribbon
General description
The MilliSentials Lab Printer Ribbon features a thermal transfer ink capable of withstanding extreme temperatures and harsh chemical solvents.
For use with the MilliSentials Lab Labeling System
Application
Laboratory Sample Labeling
Visit the Document Library
Related Content
MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.
MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.
MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.
MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.