Skip to Content
Merck
All Photos(1)

Key Documents

900401P

Avanti

pacFA

Avanti Research - A Croda Brand 900401P, powder

Synonym(s):

9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (with screw cap/argon/foil bag (900401P-1mg))

manufacturer/tradename

Avanti Research - A Croda Brand 900401P

lipid type

click reagents

shipped in

dry ice

storage temp.

-65 to -96°C

SMILES string

OC(CCCCCCCCC1(N=N1)CCCC#C)=O

InChI

1S/C15H24N2O2/c1-2-3-9-12-15(16-17-15)13-10-7-5-4-6-8-11-14(18)19/h1H,3-13H2,(H,18,19)

Application

pacFA has been used as a component for the incubation of his-tagged recombinant lysosomal integral membrane protein 2 (LIMP-2) ectodomain.

Packaging

5 mL PTFE Vial with Screw Cap/Argon/Foil Bag (900401P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Per Haberkant et al.
Biochimica et biophysica acta, 1841(8), 1022-1030 (2014-01-21)
Understanding biological processes at the mechanistic level requires a systematic charting of the physical and functional links between all cellular components. While protein-protein and protein-nucleic acid networks have been subject to many global surveys, other critical cellular components such as
PTPsigma inhibitors promote hematopoietic stem cell regeneration
Zhang Y, et al.
Nature Communications, 10(1), 1-15 (2019)
In vivo profiling and visualization of cellular protein-lipid interactions using bifunctional fatty acids.
Per Haberkant et al.
Angewandte Chemie (International ed. in English), 52(14), 4033-4038 (2013-03-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service