Skip to Content
Merck
All Photos(3)

Key Documents

841138C

Avanti

Egg PG

Avanti Research - A Croda Brand

Synonym(s):

L-α-phosphatidylglycerol (Egg, Chicken) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (841138C-25mg)
pkg of 2 × 4 mL (841138C-200mg)
pkg of 5 × 4 mL (841138C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (841138C-25mg)
25 mg/mL (841138C-200mg)
25 mg/mL (841138C-500mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI key

FJXDNGDRHUDFST-XQYKCTAGSA-M

General description

Phosphatidylglycerol (PG) is one of the major phospholipids in food and biological tissues. PG is the anionic phospholipid in the membranes of Staphylococcus sp. and Escherichia coli. PG is present in low amounts in mammalian membranes. Structurally, PG has two acyl chains esterified to a glycerol, bonded to a headgroup structure that has phosphate and carries negative charge. PG is synthesized from cytidine phosphate-diacylglycerol (CDP-DAG).

Application

Egg PG has been used:
  • as a phospholipid to test the binding ability of SEIPIN, an integral membrane protein of endoplasmic recticulum
  • as a standard for photodynamic treatment
  • as a standard for the identification of phospholipids by thin layer chromatography and electrospray ionization-mass spectra (ESI-MS)

Biochem/physiol Actions

Phosphatidylglycerol (PG) is present in amniotic fluid and indicates fetal lung maturity. It functions as a precursor for cardiolipin in animals. In plants, PG is present in thylakoid membranes and plays a central role in photosynthesis.

Packaging

5 mL Clear Glass Sealed Ampule (841138C-200mg)
5 mL Clear Glass Sealed Ampule (841138C-25mg)
5 mL Clear Glass Sealed Ampule (841138C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of an LC-MS/MS method for the quantification of choline-related compounds and phospholipids in foods and tissues
Xiong Y, et al.
Journal of Chromatography. B, Biomedical Applications, 911, 170-179 (2012)
The
An Introduction to Biological Membranes: Composition, Structure and Function, 91(3) (2016)
Photodynamic oxidation of Staphylococcus warneri membrane phospholipids: new insights based on lipidomics
Alves E, et al.
Rapid Communications in Mass Spectrometry, 27(14), 1607-1618 (2013)
The
The Membranes of Cells, 27(23) (2016)
Photodynamic oxidation of Escherichia coli membrane phospholipids: new insights based on lipidomics
Alves E, et al.
Rapid Communications in Mass Spectrometry, 27(23), 2717-2728 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service