Skip to Content
Merck
All Photos(1)

Key Documents

SML2242

Sigma-Aldrich

FTBMT

≥98% (HPLC)

Synonym(s):

4-[3-[[3-Fluoro-5-(trifluoromethyl)phenyl]methyl]-5-methyl-1H-1,2,4-triazol-1-yl]-2-methylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16F4N4O
CAS Number:
Molecular Weight:
392.35
UNSPSC Code:
12352200

Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(C1=CC=C(N2C(C)=NC(CC3=CC(F)=CC(C(F)(F)F)=C3)=N2)C=C1C)=O

InChI key

TYXSIXOYTBHZFA-UHFFFAOYSA-N

Biochem/physiol Actions

FTBMT is an orally available, selective and potent agonist of GPR52. It exhibits antipsychotic and procognitive properties without causing catalepsy in rodents. FTBMT improves recognition memory in a novel object recognition test and mitigates MK-801-induced working memory deficits in a radial arm maze test in rats.
orally available, selective and potent agonist of GPR52

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuyuki Tokumaru et al.
Bioorganic & medicinal chemistry, 25(12), 3098-3115 (2017-04-24)
G protein-coupled receptor 52 (GPR52) agonists are expected to improve the symptoms of psychiatric disorders. During exploration for a novel class of GPR52 agonists with good pharmacokinetic profiles, we synthesized 4-(3-(3-fluoro-5-(trifluoromethyl)benzyl)-5-methyl-1H-1,2,4-triazol-1-yl)-2-methylbenzamide (4u; half maximal effective concentration (EC50)=75nM, maximal response (Emax)=122%)
Keiji Nishiyama et al.
The Journal of pharmacology and experimental therapeutics, 363(2), 253-264 (2017-08-31)
GPR52 is a Gs-coupled G protein-coupled receptor that is predominantly expressed in the striatum and nucleus accumbens (NAc) and was recently proposed as a potential therapeutic target for schizophrenia. In the current study, we investigated the in vitro and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service