Skip to Content
Merck
All Photos(2)

Key Documents

D1569

Sigma-Aldrich

Anti-Dab-1 (C-terminal) from rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Dab1, Anti-Disabled homolog 1, Anti-Yotari

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

mouse, human, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 2-4 μg/mL using HEK-293T cells expressing human DAB1
western blot: 2-4 μg/mL using mouse and rat brain extracts (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DAB1(1600)
mouse ... Dab1(13131)
rat ... Dab1(266729)

General description

Disabled-1 is an intracellular adaptor protein that belongs to the Reelin signaling pathway. The amino terminus of Dab1 contains a phosphotyrosine-binding (PTB) domain. The domain interacts with NPXY sequences within the cytoplasmic regions of several membrane-bound proteins including lipoprotein receptors and amyloid precursor protein (APP) family.

Application

Anti-Dab-1 (C-terminal) was used at a working dilution of 1:2000 to probe the protein sample extracted from brain tissue of mice by western blotting in a study.

Biochem/physiol Actions

Dab1 plays a key role during brain development by controlling neuronal positioning as well as modulation of long-term potentiation (LTP) in the adult brain. Tyrosine phosphorylation of Dab1 is triggered by the binding of reelin to the lipoprotein receptors Apolipoprotein E receptor 2 (ApoER2) and the very low density lipoprotein receptor (VLDLR), thus initiating a signaling cascade that includes the Src-family kinases and Akt.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle E Whittaker et al.
The Journal of clinical investigation, 127(3), 874-887 (2017-02-07)
The mechanisms underlying the neurodevelopmental deficits associated with CHARGE syndrome, which include cerebellar hypoplasia, developmental delay, coordination problems, and autistic features, have not been identified. CHARGE syndrome has been associated with mutations in the gene encoding the ATP-dependent chromatin remodeler
Eduardo Martín-López et al.
Frontiers in neuroanatomy, 6, 15-15 (2012-06-05)
Olfaction is the most relevant chemosensory sense of the rodents. General odors are primarily detected by the main olfactory system while most pheromonal signals are received by the accessory olfactory system. The first relay in the brain occurs in the
M Trommsdorff et al.
The Journal of biological chemistry, 273(50), 33556-33560 (1998-12-05)
Apolipoprotein E, alpha2-macroglobulin, and amyloid precursor protein (APP) are involved in the development of Alzheimer's disease. All three proteins are ligands for the low density lipoprotein (LDL) receptor-related protein (LRP), an abundant neuronal surface receptor that has also been genetically
B W Howell et al.
Current biology : CB, 10(15), 877-885 (2000-08-26)
The extracellular protein Reln controls neuronal migrations in parts of the cortex, hippocampus and cerebellum. In vivo, absence of Reln correlates with up-regulation of the docking protein Dab1 and decreased Dab1 tyrosine phosphorylation. Loss of the Reln receptor proteins, apolipoprotein
Eduardo Martín-López et al.
PloS one, 6(10), e26673-e26673 (2011-11-03)
Dab1 mediates reelin signalling and plays critical roles in early brain development such as the stereotypical positioning of neurons in the brain. The olfactory bulb undergoes a prominent layering reorganization, but shows not apparent differences between wild type and reeler

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service