Skip to Content
Merck
All Photos(1)

Key Documents

Y0000675

Gemcitabine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2′-Deoxy-2′,2′-difluorocytidine; dFdC, Gemzar (Lilly), LY-188011, dFdC, dFdCyd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11F2N3O4 · HCl
CAS Number:
Molecular Weight:
299.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

gemcitabine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)C2(F)F

InChI

1S/C9H11F2N3O4.ClH/c10-9(11)6(16)4(3-15)18-7(9)14-2-1-5(12)13-8(14)17;/h1-2,4,6-7,15-16H,3H2,(H2,12,13,17);1H/t4-,6-,7-;/m1./s1

InChI key

OKKDEIYWILRZIA-OSZBKLCCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Gemcitabine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Gemcitabine is a widely used antitumor agents in both clinics and research labs. It is an antineoplastic agent and antimetabolite.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey R Infante et al.
Investigational new drugs, 33(2), 432-439 (2015-02-03)
Background This Phase 1b study aimed to determine the recommended Phase 2 dose of LY2334737, an oral pro-drug of gemcitabine, in combination with capecitabine, an oral pro-drug of 5-fluorouracil, in patients with advanced solid tumors. In addition, pharmacokinetics (PK) and
Minggen Hu et al.
Surgical endoscopy, 28(9), 2584-2591 (2014-04-08)
Laparoscopic distal splenopancreatectomy (DSP) is an effective and safe surgical modality for treating benign and borderline distal pancreatic tumors, but rarely for pancreatic cancer. This study aimed to examine the feasibility, effectiveness, and safety of laparoscopic versus laparotomic DSP in
Erez Hasnis et al.
Neoplasia (New York, N.Y.), 16(6), 501-510 (2014-06-25)
Weekly gemcitabine therapy is the major treatment offered for patients with pancreatic adenocarcinoma cancer; however, relative resistance of tumor cells to chemotherapy, rapid regrowth, and metastasis are the main causes of death within a year. Recently, the daily continuous administration
Katsumi Fukamachi et al.
PloS one, 9(10), e111481-e111481 (2014-10-28)
Pancreatic ductal adenocarcinoma (PDA) is a highly lethal disease, which is usually diagnosed in an advanced stage. We have established transgenic rats carrying a mutated K-ras gene controlled by Cre/loxP activation. The animals develop PDA which is histopathologically similar to
Rui-Qi Wang et al.
Cancer cell international, 19, 145-145 (2019-05-30)
Pancreatic cancer stem cells (CSCs), a special population of cells, renew themselves infinitely and resist to various treatment. Gramicidin A (GrA), an ionophore antibiotic derived from microorganism, can form channels across the cell membrane and disrupt cellular ionic homeostasis, leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service