Skip to Content
Merck
All Photos(2)

Key Documents

934763

Sigma-Aldrich

Iodine-doped lithium phosphorus sulfur chloride

new
greener alternative

powder, battery grade

Synonym(s):

LPSClI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li6PS5Cl0.9I0.1
Molecular Weight:
277.54
UNSPSC Code:
12352303

grade

battery grade

Quality Level

Assay

≥99% trace metals basis

form

powder

composition

Li6PS5Cl0.9I0.1

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

faint beige to dark beige

application(s)

battery manufacturing

greener alternative category

General description

Iodine-doped lithium phosphorus sulfur chloride (LPSClI) is a lithium argyrodite superionic conductor. LPSClI is a light brown-beige powder that crystallizes in a cubic crystal structure.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Sulfide-based solid electrolytes are essential components of advanced all-solid-state batteries as they exhibit the highest ionic conductivities. Argyrodite-structured sulfide solid electrolytes, like LPSClI, are among the most promising materials in this class and are currently the dominantly used solid electrolytes for all-solid-state battery fabrication. Argyrodite solid electrolytes are particularly appealing because of their ultrahigh Li-ion conductivity, quasi-stable solid–electrolyte interphase (SEI) formed with Li metal, and ability to be prepared via scalable solution-assisted synthesis approaches. The body-centered cubic-like anion framework present in LPSClI allows Li to hop directly between adjacent tetrahedral sites and facilitates the high ionic conductivity. In addition, iodine-doping improves the distribution of halogens over the sulfur sub-lattice, creating a structure with improved lithium diffusion kinetics. Because of its tailored composition, our battery grade I-doped lithium phosphorus sulfur chloride (LPSClI) is a high-performing material for solid-state battery research.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laidong Zhou et al.
Accounts of chemical research, 54(12), 2717-2728 (2021-05-26)
ConspectusAs the world transitions away from fossil energy to green and renewable energy, electrochemical energy storage increasingly becomes a vital component of the mix to conduct this transition. The central goal in developing next-generation batteries is to maximize the gravimetric
Yan Wang et al.
Nature materials, 14(10), 1026-1031 (2015-08-19)
Lithium solid electrolytes can potentially address two key limitations of the organic electrolytes used in today's lithium-ion batteries, namely, their flammability and limited electrochemical stability. However, achieving a Li(+) conductivity in the solid state comparable to existing liquid electrolytes (>1 mS cm(-1))
An argyrodite sulfide-based superionic conductor synthesized by a liquid-phase technique with tetrahydrofuran and ethanol
So Y, et al.
Journal of Material Chemistry A, 7, 558-566 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service