Skip to Content
Merck
All Photos(2)

Documents

182877

Sigma-Aldrich

Polystyrene-block-polybutadiene-block-polystyrene

styrene 30 wt. %, average Mw ~140,000 by GPC, contains <0.5 wt. % antioxidant

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x (CH2CH=CHCH2)y [CH2CH(C6H5)]x
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

chunks

Quality Level

mol wt

average Mw ~140,000 by GPC

contains

<0.5 wt. % antioxidant

composition

styrene, 30 wt. %

intrinsic viscosity

1.0(lit.)

density

0.94 g/mL at 25 °C (lit.)

InChI

1S/C8H8.C4H6/c1-2-8-6-4-3-5-7-8;1-3-4-2/h2-7H,1H2;3-4H,1-2H2

InChI key

MTAZNLWOLGHBHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Polystyrene-block-Polybutadiene-block-Polystyrene Triblock Copolymer Meets Silica: From Modification of Copolymer to Formation of Mesoporous Silica: Describes the preparation of mesoporous silica materials assisted by a PS-b-PB-b-PS triblock copolymer. This work involves functionalizing the copolymer with (3-mercaptopropyl) triethoxysilane via a thiol-ene radical addition, which is then employed to perform sol–gel reactions with tetraethoxysilane, providing insights into the multifunctional capabilities of these materials (R Yu, S Zheng, 2015).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service