Skip to Content
Merck
All Photos(1)

Documents

35605

Sigma-Aldrich

(−)-(8,8-Dichlorocamphorylsulfonyl)oxaziridine

≥95.0% (T)

Synonym(s):

(−)-3,3-Dichloro-2,N-epoxy-exo-10,2-bornanesultam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13Cl2NO3S
CAS Number:
Molecular Weight:
298.19
Beilstein:
3653351
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (T)

optical activity

[α]20/D −91±3°, c = 0.5% in chloroform

storage temp.

2-8°C

SMILES string

CC1(C)[C@]2([H])C(Cl)(Cl)[C@@](N3S(C4)(=O)=O)(O3)[C@@]14CC2

InChI

1S/C10H13Cl2NO3S/c1-7(2)6-3-4-8(7)5-17(14,15)13-10(8,16-13)9(6,11)12/h6H,3-5H2,1-2H3/t6-,8-,10+,13?/m1/s1

InChI key

HAMBQYFZDBYWHU-VYBWYVTGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pamela J Weathers et al.
Industrial crops and products, 62, 173-178 (2014-09-18)
Artemisia annua L., long used as a tea infusion in traditional Chinese medicine, produces artemisinin. Although artemisinin is currently used as artemisinin-based combination therapy (ACT) against malaria, oral consumption of dried leaves from the plant showed efficacy and will be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service