Skip to Content
Merck
All Photos(2)

Documents

345350

Sigma-Aldrich

(1S)-(+)-(10-Camphorsulfonyl)oxaziridine

Synonym(s):

(1S)-(+)-(Camphorylsulfonyl)oxaziridine, (1S)-(+)-2,N-Epoxy-exo-10,2-bornanesultam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO3S
CAS Number:
Molecular Weight:
229.30
Beilstein:
6274370
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

optical activity

[α]28/D +45°, c = 2 in chloroform

impurities

<1% toluene

mp

172-174 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H]2CC[C@]13CS(=O)(=O)N4O[C@@]34C2

InChI

1S/C10H15NO3S/c1-8(2)7-3-4-9(8)6-15(12,13)11-10(9,5-7)14-11/h7H,3-6H2,1-2H3/t7-,9+,10+,11?/m1/s1

InChI key

GBBJBUGPGFNISJ-YDQXZVTASA-N

Looking for similar products? Visit Product Comparison Guide

Application

(1S)-(+)-(10-Camphorsulfonyl)oxaziridine can be used:
  • To convert prochiral ketone enolates into optically active α-hydroxy ketones via enantioselective asymmetric oxidation.
  • In the synthesis of thymidine oligonucleotides connected through pyrophosphates.
  • In the asymmetric synthesis of proton pump inhibitors like (R)-Rabeprazole sodium and (R)-Lansoprazole sodium from the corresponding DBU salt of prochiral sulfide.
  • In the preparation of phosphonoacetate and thiophosphonoacetate oligodeoxynucleotides by oxidizing the corresponding phosphinoacetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the synthesis of oligonucleotides interconnected through pyrophosphate linkages
Kistemaker HAV, et al.
European Journal of Organic Chemistry, 2015(27), 6084-6091 (2015)
Chemistry of oxaziridines. 14. Asymmetric oxidation of ketone enolates using enantiomerically pure (camphorylsulfonyl) oxaziridine
Davis FA, et al.
Journal of the American Chemical Society, 112(18), 6679-6690 (1990)
Davis oxaziridine-mediated asymmetric synthesis of proton pump inhibitors using DBU salt of prochiral sulfide
Mahale RD, et al.
Organic Process Research & Development, 14(5), 1264-1268 (2010)
Douglas J Dellinger et al.
Journal of the American Chemical Society, 125(4), 940-950 (2003-01-23)
Phosphonoacetate and thiophosphonoacetate oligodeoxynucleotides were prepared via a solid-phase synthesis strategy. Under Reformatsky reaction conditions, novel esterified acetic acid phosphinodiamidites were synthesized and condensed with appropriately protected 5'-O-(4, 4'-dimethoxytrityl)-2'-deoxynucleosides to yield 3'-O-phosphinoamidite reactive monomers. These synthons when activated with tetrazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service