Z693707
Duran® filtering flask, with Keck® assembly set
capacity 500 mL
Synonym(s):
212044453
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
flat bottom flask
Joint
joint ST/NS neck34 mm I.D.
sterility
non-sterile
packaging
pkg of 1 ea
pkg of 10 ea
manufacturer/tradename
DURAN® 212044453
availability
not available in USA and Canada
capacity
500 mL
General description
The Duran filtering flask includes a Keck clip assembly and is idea for vacuum filtrations. The plastic hose connections meet safety and product regulations, and the flask has the Erlenmeyer shape.
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Keck is a registered trademark of Adcuram Lab Technologies GmbH