DWK211949157
Duran® Filtering Flasks
Bottle shape, WITH KECK ASSEMBLY SET
Synonym(s):
211949157
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 211949157
capacity
20000 mL
Looking for similar products? Visit Product Comparison Guide
General description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. Büchner filter flasks are used in conjunction with Büchner filter funnels and other types of funnels in vacuum filtration applications.
Application
Separations by vacuum filtration, Filtration at reduced pressure, Filtrieren, Vacuum filtration, Safe working under vacuum, Boro 3.3, PBT, PP, VMQ, EPDM
Features and Benefits
Vacuum resistant due to the heavy wall thickness; These filtering flasks fulfil the regulations of the equipment and product safety regulations; Fired-on, highly durable white print; Fired-on, highly durable white print;
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service