Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502973

Sigma-Aldrich

Anti-TIMP3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Metalloproteinase inhibitor 3, Protein MIG-5, TIMP-3, Tissue inhibitor of metalloproteinases 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 24 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMP3(7078)

Related Categories

General description

Tissue inhibitor of metalloproteinases 3 (TIMP3) belongs to the matrix metalloproteinases inhibitor family. TIMP3 gene is mapped to human chromosome 22q12.3. TIMP3 possesses a C-terminal N-linked glycosylation site at asparagine residue. This protein comprises 12 conserved cysteine residues stabilized by six disulfide bonds in its secondary structure. TIMP-3 gene expression is higher in murine lung and kidney.

Immunogen

The antiserum was produced against synthesized peptide derived from human TIMP3.

Immunogen Range: 91-140

Biochem/physiol Actions

Tissue inhibitor of metalloproteinases 3 (TIMP3) inhibits membrane-type matrix metalloproteinases (MT-MMPs) and the tumor necrosis factor α–converting enzyme (TACE). It displays a high affinity for the proteoglycans associated with extracellular matrix (ECM) and displays tight binding with sulfated glycosaminoglycans. Mutations in the TIMP-3 gene is implicated in Sorsby′s fundus dystrophy which is characterized by choroidal neovascularization, vision loss, and abnormal Bruch′s membrane thickening. Downregulation of TIMP3 gene due to allelic losses on 22q12 chromosomal location is correlated to meningiomas. TIMP3 participates in cardiac remodeling. Low levels of TIMP3 protein are observed in scenarios of ischemic and dilated cardiomyopathy. This protein elicits proapoptotic activity and is an inhibitor of angiogenesis and tumor growth.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne M Macgregor et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 57(3), 207-213 (2008-10-29)
Tissue inhibitor of matrix metalloproteinase-3 (TIMP-3) is an important regulator of matrix metalloproteinase activity in many types of disease, including atherosclerosis, neoplasia, and inflammatory conditions. Among TIMPs, TIMP-3 uniquely binds the extracellular matrix (ECM). We performed IHC staining on 17
Jian Hua Qi et al.
Nature medicine, 9(4), 407-415 (2003-03-26)
Tissue inhibitor of metalloproteinases-3 (TIMP3) is one of four members of a family of proteins that were originally classified according to their ability to inhibit matrix metalloproteinases (MMP). TIMP3, which encodes a potent angiogenesis inhibitor, is mutated in Sorsby fundus
Robert Visse et al.
Circulation research, 92(8), 827-839 (2003-05-06)
Matrix metalloproteinases (MMPs), also designated matrixins, hydrolyze components of the extracellular matrix. These proteinases play a central role in many biological processes, such as embryogenesis, normal tissue remodeling, wound healing, and angiogenesis, and in diseases such as atheroma, arthritis, cancer
Dong Fan et al.
Frontiers in physiology, 11, 661-661 (2020-07-03)
Tissue inhibitor of metalloproteinase 3 (TIMP3) is unique among the four TIMPs due to its extracellular matrix (ECM)-binding property and broad range of inhibitory substrates that includes matrix metalloproteinases (MMPs), a disintegrin and metalloproteinases (ADAMs), and ADAM with thrombospondin motifs
Dimitri Barski et al.
Brain pathology (Zurich, Switzerland), 20(3), 623-631 (2009-11-20)
The gene for the tissue inhibitor of metalloproteinase 3 (TIMP3) on 22q12.3 had been reported to be inactivated by promoter methylation in various types of cancers, with controversial findings in meningiomas. We performed direct sodium bisulfite sequencing in a series

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service