Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2131

Sigma-Aldrich

Anti-BMAL1/ARNTL Antibody, clone 1J12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Aryl hydrocarbon receptor nuclear translocator-like protein 1;Basic-helix-loop-helix-PAS protein MOP3;Brain and muscle ARNT-like 1;Class E basic helix-loop-helix protein 5;bHLHe5, Member of PAS protein 3;PAS domain-containing protein 3;bHLH-PAS protein JA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J12, recombinant monoclonal

description

1J12 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 68.76 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ARNTL(406)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J12 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects BMAL1/ARNTL. It targets an epitope within 19 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of human BMAL1/ARNTL.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected BMAL1/ARNTL in NIH3T3 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected recombinant human BMAL1/ARNTL

Enzyme Immunoassay (ELISA) Analysis: Various dilutions from a representative lot detected ABP and His tagged recombinant human BMAL1/ARNTL

Affinity Binding Assay: A representative lot of this antibody bound BMAL1/ARNTL peptide with a KD of 5.8 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Aryl hydrocarbon receptor nuclear translocator-like protein 1 (UniProt: O00327; also known as Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, bHLHe5, Member of PAS protein 3, PAS domain-containing protein 3, bHLH-PAS protein JAP3) is encoded by the ARNTL (also known as BHLHE5, BMAL1, MOP3, PASD3) gene (Gene ID: 406) in human. BMAL1 is a transcriptional activator that forms a core component of the circadian clock. These components include ARNTL/BMAL1, CLOCK, NPAS2, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1, and CRY2.  CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2 form the positive limb of the feedback loop. They act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes that are involved in key metabolic processes. PER1/2/3 and CRY1/2, act as transcriptional repressors and form the negative limb of the feedback loop. They interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity. BMLA1 is reported to be phosphorylated upon dimerization with CLOCK and this phosphorylation enhances its transcriptional activity and reduces the stability of the CLOCK-ARNTL/BMAL1 heterodimer by promoting its degradation. BMAL1 is shown to positively regulate myogenesis and negatively regulate adipogenesis via the transcriptional control of the genes of the canonical Wnt signaling pathway. BMAL1 is the only component of the mammalian circadian clock whose deletion in a murine model is shown to generate arrhythmicity. BMAL1 null-mice are reported to have reproductive problems, reduced locomotor activity, and premature aging. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, Z., et al. (2013). Cell Res. 23(2); 213-224).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service