Skip to Content
Merck
All Photos(2)

Key Documents

AV43935

Sigma-Aldrich

Anti-SLC16A8 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-MCT3, Anti-Solute carrier family 16, member 8 (monocarboxylic acid transporter 3)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

guinea pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC16A8(23539)

Immunogen

Synthetic peptide directed towards the middle region of human SLC16A8

Application

Anti-SLC16A8 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC16A8 (MCT3) is a proton-coupled monocarboxylate transporter that facilitates the movement of lactate across the cell membranes. Mutation in the gene for MCT3 results in altered visual function in mice and has been associated with age-related macular degeneration.

Sequence

Synthetic peptide located within the following region: RAFAVYAVTKFLMALGLFVPAILLVNYAKDAGVPDTDAAFLLSIVGFVDI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars G Fritsche et al.
Nature genetics, 45(4), 433-439 (2013-03-05)
Age-related macular degeneration (AMD) is a common cause of blindness in older individuals. To accelerate the understanding of AMD biology and help design new therapies, we executed a collaborative genome-wide association study, including >17,100 advanced AMD cases and >60,000 controls
H Yoon et al.
Genomics, 60(3), 366-370 (1999-09-24)
Lactate transport across cell membranes is mediated by a family of proton-coupled monocarboxylate transporters (MCTs). The retinal pigment epithelium (RPE) expresses a unique member of this family, MCT3. A portion of the human MCT3 gene was cloned by polymerase chain
Timothy A Blenkinsop et al.
Investigative ophthalmology & visual science, 56(12), 7085-7099 (2015-11-06)
We tested what native features have been preserved with a new culture protocol for adult human RPE. We cultured RPE from adult human eyes. Standard protocols for immunohistochemistry, electron microscopy, electrophysiology, fluid transport, and ELISA were used. Confluent monolayers of
Lauren L Daniele et al.
American journal of physiology. Cell physiology, 295(2), C451-C457 (2008-06-06)
To meet the high-energy demands of photoreceptor cells, the outer retina metabolizes glucose through glycolytic and oxidative pathways, resulting in large-scale production of lactate and CO(2). Mct3, a proton-coupled monocarboxylate transporter, is critically positioned to facilitate transport of lactate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service