Skip to Content
Merck
All Photos(1)

Key Documents

W407815

Sigma-Aldrich

N-Benzoylanthranilic acid

≥97%, FG

Synonym(s):

2-(Benzoylamino)benzoic acid, Dianthramid B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NO3
CAS Number:
Molecular Weight:
241.24
FEMA Number:
4078
MDL number:
UNSPSC Code:
12352106
Flavis number:
16.087
NACRES:
NA.21
Organoleptic:
fruity
grade:
FG
Agency:
follows IFRA guidelines
food allergen:
no known allergens

grade

FG

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU 1223/2009
EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity

SMILES string

N(c2c(cccc2)C(=O)O)C(=O)c1ccccc1

InChI

1S/C14H11NO3/c16-13(10-6-2-1-3-7-10)15-12-9-5-4-8-11(12)14(17)18/h1-9H,(H,15,16)(H,17,18)

InChI key

WXVLIIDDWFGYCV-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service