Skip to Content
Merck
All Photos(1)

Key Documents

NIST2270

Perdeuterated PAH-II solution in hexane/toluene

NIST® SRM® 2270

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

format

multi-component solution

General description

This Standard Reference Material (SRM) is intended primarily for use as an internal standard, or surrogate internal standard, solution that is used to spike both the unknown sample and the calibration or external standard solution of non-labeled PAHs. A unit of SRM 2270 consists of five 2 mL ampoules, each containing approximately 1.2 mL of a solution of six perdeuterated polycyclic aromatic hydrocarbons (PAHs) in hexane/toluene (96:4 volume fraction). For more information, please refer to the COA and SDS.

SRM 2270_cert

SRM 2270_SDS

Features and Benefits

  • NIST certified concentration values of six perdeuterated PAHs are reported based on the results obtained from the gravimetric preparation of the solution and the analytical results determined by using gas chromatography.
  • Information Values for Perdeuterated PAHs are provided.
  • Maintenance and expiration of SRM certification, instructions for handling, storage, and use are provided on NIST certificate.

Other Notes

Certified for the analytes listed below.
Naphthalene-d8 (C10H8), Acenaphthene-d10 (C12H10), Pyrene-d10 (C16H10), Benzo[a]pyrene-d12 (C20H12), Perylene-d12 (C20H12), Benzo[ghi]perylene-d12 (C22H12)

Reference/Informational value is provided for the following analytes.
Naphthalene-d8 (C10H8), Acenaphthene-d10 (C12H10), Pyrene-d10 (C16H10), Benzo[a]pyrene-d12 (C20H12), Perylene-d12 (C20H12), Benzo[ghi]perylene-d12 (C22H12)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service