There isn't a Purge and Trap System specifically optimized for the release of acrolein. However, it seems that propyl gallate can be used to effectively eliminate acrolein at high temperatures. An example of an applicable product would be 48710-100G-F.
28994-U
Purge/Trap K
Vocarb® 3000, configured for StableFlex™ Stratum
Synonym(s):
VOCARB purge trap (Tekmar)
About This Item
Recommended Products
Product Name
Purge/Trap K Vocarb® 3000, configured for Tekmar® Stratum
application(s)
industrial hygiene
Quality Level
compatibility
configured for Tekmar® Stratum
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is there a Purge/Trap system that is optimized for the elimination of acrolein?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
Are the Supelco Purge Traps for Tekmar Stratum compatible with the Tekmar Atomx?
1 answer-
Purge traps manufactured for the Tekmar Stratum PTC Purge and Trap Concentrator are the same traps used for the Tekmar Atomx Automated Sample Prep System.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service