Skip to Content
Merck
All Photos(1)

Key Documents

8.52310

Sigma-Aldrich

Fmoc-SDMA(Boc)2-ONa

Novabiochem®

Synonym(s):

Fmoc-SDMA(Boc)2-ONa, N-α-Fmoc-N,NÆ-δ-dimethyl-N,NÆ-δ-di-t.-butoxycarbonyl-L-arginine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H43N4NaO8
Molecular Weight:
646.71
UNSPSC Code:
12352209
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

Assay

≥90.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

-10 to -25°C

General description

A derivative for the introduction of symmetric dimethyl-arginine during Fmoc SPPS. Coupling can be carried out using any standard activation method. Removal of the Boc protecting groups occurs during the course of the TFA-mediated cleavage reaction. Ref [1] contains methods and protocols for the synthesis of arrays of histone-related peptides containing methylated arginine and lysine-residues.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS

Literature references

[1] S. Rothbart, et al. (2012) Methods Enzymol., 512, 107.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 90.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.

Protocols

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service