Skip to Content
Merck
All Photos(1)

Documents

8.22122

Sigma-Aldrich

Phenanthrene

for synthesis

Synonym(s):

Phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352005
EC Index Number:
201-581-5
NACRES:
NA.22

vapor pressure

0.00012 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

autoignition temp.

>450 °C

potency

1800 mg/kg LD50, oral (Rat)

bp

336 °C/1013 hPa

mp

97-100 °C

transition temp

flash point 171 °C

solubility

1.15 mg/L

bulk density

690 kg/m3

storage temp.

2-30°C

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H

InChI key

YNPNZTXNASCQKK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 97 °C
Melting range (upper value): ≤ 100 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service