Skip to Content
Merck
All Photos(2)

Documents

695114

Sigma-Aldrich

Phenanthrene

sublimed grade, ≥99.5%

Synonym(s):

Ravatite, [3]Helicene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Beilstein:
1905428
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Quality Level

Assay

≥99.5%

form

crystals
powder and chunks
tubes

bp

340 °C (lit.)

mp

98-100 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

c1ccc2c(c1)ccc3ccccc23

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H

InChI key

YNPNZTXNASCQKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenanthrene is a polycyclic aromatic hydrocarbon (PAH) which consists of three benzene rings that are prepared by Bardhan-Sengupta phenanthrene synthesis. Its properties include low volatility, high toxicity and resistance to microbial biodegradation.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bardhan-Sengupta Synthesis
Comprehensive Organic Name Reactions and Reagents (2010)
Phenanthrene (2012)
Biodegradation of phenanthrene in river sediment
Yuan SY, et al.
Chemosphere, 43(3), 273-278 (2001)
Jennifer R McKelvie et al.
Environmental pollution (Barking, Essex : 1987), 175, 75-81 (2013-01-23)
We evaluated the correlation between soil organic carbon (OC) content and metabolic responses of Eisenia fetida earthworms after exposure to phenanthrene (58 ± 3 mg/kg) spiked into seven artificial soils with OC contents ranging from 1 to 27% OC. Principal
Raïssa Kom Regonne et al.
Environmental pollution (Barking, Essex : 1987), 180, 145-151 (2013-06-19)
Efficient bioremediation of PAH-contaminated sites is limited by the hydrophobic character and poor bioavailability of pollutants. In this study, stable isotope probing (SIP) was implemented to track bacteria that can degrade PAHs adsorbed on hydrophobic sorbents. Temperate and tropical soils

Articles

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service