104309
L-Methionine sulfoximine
EMPROVE® ESSENTIAL
Pharma Manufacturing
Synonym(s):
L-S-(3-Amino-3-carboxypropyl)-S-methylsulfoximine
About This Item
Recommended Products
product line
EMPROVE® ESSENTIAL
Quality Level
form
solid
packaging
pkg of 10g (1043090010)
pkg of 1g (1043090001)
pkg of 5g (1043090005)
pkg of
mp
>210 °C (dec.) (lit.)
suitability
suitable for manufacturing use
SMILES string
CS(=N)(=O)CC[C@H](N)C(O)=O
InChI
1S/C5H12N2O3S/c1-11(7,10)3-2-4(6)5(8)9/h4,7H,2-3,6H2,1H3,(H,8,9)/t4-,11?/m0/s1
InChI key
SXTAYKAGBXMACB-DPVSGNNYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
M-Clarity Program
As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.
Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
Legal Information
replaced by
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service