Skip to Content
Merck
All Photos(1)

Key Documents

14365C

SAFC

EX-CELL® CD CHO Fusion

Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

sterility

sterile-filtered

product line

EX-CELL®

form

liquid

technique(s)

cell culture | mammalian: suitable (suspension)

components

L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EX-CELL® CD CHO Fusion is a chemically defined, animal-component free medium developed for the long-term growth of Chinese Hamster Ovary (CHO) cells. The absence of any large macro molecules allows for isolation and purification of secreted proteins from the cells. This medium is supplied without L-glutamine to aid in media stability, to avoid L-glutamine degradation that causes ammonia build-up and to provide an appropriate medium for the culture of CHO cells using glutamine synthetase selection (example: MilliporeSigma’s CHOZN GS knockout cell line). This medium does not contain hypoxanthine or thymidine to allow for its use with dihydrofolate reductase(DHFR-) gene amplification systems (example: MilliporeSigma’s CHOZN DHFR knockout cell line).

Quantity

Formulated to contain 20.9 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noriko Yamano-Adachi et al.
Scientific reports, 10(1), 17612-17612 (2020-10-21)
Chinese hamster (Cricetulus griseus) ovary-derived Chinese hamster ovary (CHO) cells are the most commonly used mammalian hosts for the industrial production of recombinant therapeutics because of their ability to fold, assemble, and perform post-translational modifications, such as glycosylation, on proteins.
Qiong Wang et al.
Biotechnology and bioengineering, 116(9), 2303-2315 (2019-05-08)
With the increasing demand to provide more detailed quality attributes, more sophisticated glycan analysis tools are highly desirable for biopharmaceutical manufacturing. Here, we performed an intact glycopeptide analysis method to simultaneously analyze the site-specific N- and O-glycan profiles of the
Weiyi Liu et al.
Biotechnology progress, 37(1), e3061-e3061 (2020-08-05)
Antibody-dependent cellular cytotoxicity (ADCC) is the primary mechanism of actions for several marketed therapeutic antibodies (mAbs) and for many more in clinical trials. The ADCC efficacy is highly dependent on the ability of therapeutic mAbs to recruit effector cells such

Articles

This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.

This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.

This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.

This page describes the financial and operational advantages offered by the Cellicon® Cell Retention Device in an N-1 perfusion process at the 2,000 L scale in comparison to a cell retention device from another supplier.

See All

Related Content

Eliminate the need to clean, autoclave and assemble your traditional glass bioreactors and save days of downtime with the Mobius® 3L Single-use Bioreactor.

Learn more about cell line development and the relevant products you need for this process.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service