Key Documents
SYNM531042
Synthware nitrogen/argon line manifold with PTFE stopcocks
stopcock bore 2 mm, Ports: 4, Hose Connections: left
Synonym(s):
Synthware M531042, vacuum manifold, gas manifold
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
manufacturer/tradename
M531042
stopcock bore
2 mm
General description
Synthware glass manifold featuring high vacuum hollow glass stopcocks, the smaller O.D. 17 mm front tube is designed for inert gases, while the larger O.D. 22 mm back tube is for vacuum, which has a 35/20 socket on the left-hand side. Hose connections (10 mm) are located front-left-right.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)