Key Documents
SYNM490064
Synthware vacuum manifold with PTFE stopcocks
number of ports, 6, Hose Connection: left-right, stopcock size 4 mm
Synonym(s):
Synthware M490064, vaccum unit
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Pricing and availability is not currently available.
feature
number of ports 6
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M490064
stopcock size
4 mm
General description
General purpose manifold with 2 mm or 4 mm PTFE stopcocks. Main tube is 22 mm O.D. with serrated hose connection(10 mm O.D.) at both ends. Delivery ports have 10 mm serrated hose connections. Distance between stopcocks is75 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)