GF63565599
Tungsten-rhenium alloy, W97Re3
foil, 0.2m coil, .5mm coil width, 0.05mm thickness, annealed
Synonym(s):
W 040250
Select a Size
Select a Size
About This Item
Recommended Products
form
annealed
foil (coil)
manufacturer/tradename
Goodfellow 635-655-99
L × W × thickness
2 m × 0.5 mm × 0.05 mm
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service