Skip to Content
BG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten-rhenium alloy, W97Re3
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 57
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF95295448
Tungsten-rhenium alloy, W97Re3,
1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF69744213
Tungsten-rhenium alloy, W97Re3,
foil, 0.1m coil, .5mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF45699047
Tungsten-rhenium alloy, W97Re3,
foil, 0.5m coil, .5mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF86362364
Tungsten-rhenium alloy, W97Re3,
0.5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF75297684
Tungsten-rhenium alloy, W97Re3,
20m wire, 0.02mm diameter, annealed
Expand
View Pricing
GF85481499
Tungsten-rhenium alloy, W97Re3,
50m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF17783860
Tungsten-rhenium alloy, W97Re3,
foil, 0.05m coil, .5mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF56996072
Tungsten-rhenium alloy, W97Re3,
20m wire, 0.2mm diameter, annealed
Expand
View Pricing
GF95043999
Tungsten-rhenium alloy, W97Re3,
20m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF63565599
Tungsten-rhenium alloy, W97Re3,
foil, 0.2m coil, .5mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF48288055
Tungsten-rhenium alloy, W97Re3,
2m wire, 0.2mm diameter, annealed
Expand
View Pricing
GF84870401
Tungsten-rhenium alloy, W97Re3,
1m wire, 0.15mm diameter, annealed
Expand
View Pricing
GF86851020
Tungsten-rhenium alloy, W97Re3,
1m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF49490893
Tungsten-rhenium alloy, W97Re3,
0.1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF87303966
Tungsten-rhenium alloy, W97Re3,
10m wire, 0.15mm diameter, annealed
Expand
View Pricing
GF15999801
Tungsten-rhenium alloy, W97Re3,
20m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF65649066
Tungsten-rhenium alloy, W97Re3,
100m wire, 0.01mm diameter, as drawn
Expand
View Pricing
GF27005374
Tungsten-rhenium alloy, W97Re3,
10m wire, 0.01mm diameter, as drawn
Expand
View Pricing
GF99094809
Tungsten-rhenium alloy, W97Re3,
0.5m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF36006689
Tungsten-rhenium alloy, W97Re3,
2m wire, 0.25mm diameter, annealed
Expand
View Pricing
Page 1 of 3
Page 1 of 3