Skip to Content
Merck
All Photos(1)

Documents

518972

Sigma-Aldrich

trans-2-(4-Fluorophenyl)vinylboronic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH=CHB(OH)2
CAS Number:
Molecular Weight:
165.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

182-186 °C (lit.)

SMILES string

OB(O)\C=C\c1ccc(F)cc1

InChI

1S/C8H8BFO2/c10-8-3-1-7(2-4-8)5-6-9(11)12/h1-6,11-12H/b6-5+

InChI key

GBNJRIQSFJFDII-AATRIKPKSA-N

Application

Reactant for:
  • Suzuki-Miyaura coupling reactions
  • Preparation of biologically and pharmacologically active molecules
  • Conjugate addition reactions

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service