519022
trans-2-[4-(Trifluoromethyl)phenyl]vinylboronic acid
≥95%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H4CF3CHCHB(OH)2
CAS Number:
Molecular Weight:
215.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
solid
mp
204-210 °C (lit.)
functional group
fluoro
SMILES string
[H]\C(B(O)O)=C(\[H])c1ccc(cc1)C(F)(F)F
InChI
1S/C9H8BF3O2/c11-9(12,13)8-3-1-7(2-4-8)5-6-10(14)15/h1-6,14-15H/b6-5+
InChI key
BBNQFBHQOPZKTI-AATRIKPKSA-N
Application
Reactant for:
Microwave-assisted Suzuki-Miyaura cross-coupling
Cobalt-catalyzed coupling reactions
Preparation of vinylic MIDA boronates
Synthesis of C2-aryl pyrrolobenzodiazepine antitumor agents
Suzuki-Miyaura cross-coupling reaction
Microwave-assisted Suzuki-Miyaura cross-coupling
Cobalt-catalyzed coupling reactions
Preparation of vinylic MIDA boronates
Synthesis of C2-aryl pyrrolobenzodiazepine antitumor agents
Suzuki-Miyaura cross-coupling reaction
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service