Skip to Content
Merck
All Photos(1)

Key Documents

1711155

USP

Vecuronium bromide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-[(2β,3α,5α,16β,17β)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-piperidinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H57BrN2O4
CAS Number:
Molecular Weight:
637.73
Beilstein:
4837308
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vecuronium

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(C)=O)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1N6CCCCC6

InChI

1S/C34H57N2O4.BrH/c1-23(37)39-31-20-25-12-13-26-27(34(25,4)22-29(31)35-16-8-6-9-17-35)14-15-33(3)28(26)21-30(32(33)40-24(2)38)36(5)18-10-7-11-19-36;/h25-32H,6-22H2,1-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1

InChI key

VEPSYABRBFXYIB-PWXDFCLTSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vecuronium bromide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Pancuronium Bromide
  • Pancuronium Bromide Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Bi et al.
Journal of cellular and molecular medicine, 18(6), 1226-1235 (2014-03-22)
Preservation or restoration of normal alveolar epithelial barrier function is crucial for pulmonary oedema resolution. Keratinocyte growth factor-2 (KGF-2), a potent epithelial cell mitogen, may have a role in preventing ventilator-induced lung injury (VILI), which occurs frequently in mechanically ventilated
Dimitri Tränkner et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(31), 11515-11520 (2014-07-23)
Asthma is a common debilitating inflammatory lung disease affecting over 200 million people worldwide. Here, we investigated neurogenic components involved in asthmatic-like attacks using the ovalbumin-sensitized murine model of the disease, and identified a specific population of neurons that are
Koichiro Shinozaki et al.
The Journal of surgical research, 195(1), 196-203 (2015-01-04)
Emerging therapies for prolonged cardiac arrest (CA) include advanced circulatory interventions like emergency cardiopulmonary bypass (ECPB) and continuous venovenous hemofiltration (CVVHF). However, preclinical studies are limited because of the absence of a practical method of using CVVHF along with ECPB
Ana P Abdala et al.
American journal of respiratory cell and molecular biology, 50(6), 1031-1039 (2013-12-20)
Disturbances in respiration are common and debilitating features of Rett syndrome (RTT). A previous study showed that the 5-HT1a receptor agonist (R)-(+)-8-hydroxy-dipropyl-2-aminotetralin hydrobromide (8-OH-DPAT) significantly reduced the incidence of apnea and the irregular breathing pattern in a mouse model of
Luciana Mesquita Passamani et al.
PloS one, 9(7), e101886-e101886 (2014-07-10)
Our aim was to assess the timing and mechanisms of the sympathoexcitation that occurs immediately after coronary ligation. We recorded thoracic sympathetic (tSNA) and phrenic activities, heart rate (HR) and perfusion pressure in Wistar rats subjected to either ligation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service