Skip to Content
Merck
All Photos(1)

Key Documents

SML2168

Sigma-Aldrich

TAK-475

≥98% (HPLC)

Synonym(s):

1-[2-[(3R,5S)-1-[3-(Acetyloxy)-2,2-dimethylpropyl]-7-chloro-5-(2,3-dimethoxyphenyl)-1,2,3,5-tetrahydro-2-oxo-4,1-benzoxazepin-3-yl]acetyl]-4-piperidineacetic acid, Lapaquistat acetate, TAK 475

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H41ClN2O9
CAS Number:
Molecular Weight:
645.14
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1[C@@H](CC(N2CCC(CC(O)=O)CC2)=O)O[C@H](C3=CC=CC(OC)=C3OC)C4=C(C=CC(Cl)=C4)N1CC(C)(C)COC(C)=O

InChI

1S/C33H41ClN2O9/c1-20(37)44-19-33(2,3)18-36-25-10-9-22(34)16-24(25)30(23-7-6-8-26(42-4)31(23)43-5)45-27(32(36)41)17-28(38)35-13-11-21(12-14-35)15-29(39)40/h6-10,16,21,27,30H,11-15,17-19H2,1-5H3,(H,39,40)/t27-,30-/m1/s1

InChI key

CMLUGNQVANVZHY-POURPWNDSA-N

Application

TAK-475 has been used:
  • as a farnesyl-diphosphate farnesyltransferase 1 (FDFT1) inhibitor to study its effects on cell signaling in pancreatic ductal adenocarcinoma cells
  • as a squalene synthase (SQS) inhibitor to stabilize HMG-CoA reductase (HMGCR)-dCat-ELuc proteins
  • as an SQS inhibitor to study its effects on the migration of prostate cancer cells

Biochem/physiol Actions

TAK-475 (Lapaquistat acetate) is a potent and selective squalene synthase inhibitor. TAK-475 effectively lowers low-density lipoprotein cholesterol in human. Clinical development of TAK-475 was discontinued due to hepatotoxicity seen in two patients receiving a high dose.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ebihara et al.
Drug research, 66(6), 287-292 (2016-02-04)
The pharmacokinetics of TAK-475 (lapaquistat acetate), a squalene synthase inhibitor, was investigated in rats and dogs. After oral administration of (14)C-labeled TAK-475 ([(14)C]TAK-475) to rats and dogs at a dose of 10 mg/kg, the bioavailability (BA) was relatively low at 3.5
Evan A Stein et al.
Circulation, 123(18), 1974-1985 (2011-04-27)
Lapaquistat acetate is a squalene synthase inhibitor investigated for the treatment of hypercholesterolemia. This report summarizes the phase 2 and 3 results from the lapaquistat clinical program, which was halted at an advanced stage as a result of potential hepatic
Chao Huang et al.
Journal of extracellular vesicles, 9(1), 1692417-1692417 (2019-12-07)
Tumour metastasis suppressor KAI1/CD82 inhibits tumour cell movement. As a transmembrane protein, tetraspanin CD82 bridges the interactions between membrane microdomains of lipid rafts and tetraspanin-enriched microdomains (TEMs). In this study, we found that CD82 and other tetraspanins contain cholesterol recognition/interaction
Nobutaka Suzuki et al.
SpringerPlus, 5(1), 1429-1429 (2016-09-22)
TAK-475 (lapaquistat acetate) and its active metabolite-I (TAK-475 M-I) inhibit squalene synthase, which catalyzes the conversion of farnesyl diphosphate (FPP) to squalene. FPP is a substrate for synthesis of other mevalonate-derived isoprenoids (MDIs) such as farnesol (FOH), geranlygeranyl diphosphate (GGPP)
Ikuya Sagimori et al.
Bioorganic & medicinal chemistry, 28(3), 115298-115298 (2020-01-07)
HMG-CoA reductase (HMGCR) is the rate-limiting enzyme in the cholesterol biosynthetic pathway, and is the target of cholesterol-lowering drugs, statins. Previous studies have demonstrated that the enzyme activity is regulated by sterol-induced degradation in addition to transcriptional regulation through sterol-regulatory-element-binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service